Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

288578

Supelco

Silica gel

5-25 μm avg. part. size, without binder, average pore diameter 60 Å, pore volume 0.75 cm3/g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200

contains

fluorescent indicator

impurities

≤0.001% Fe3+

avg. part. size

5-25 μm

pore size

0.75 cm3/g pore volume
60 Å average pore diameter

pH

~6.8 (10% in aq. suspension)

bp

2230 °C

mp

>1600 °C

anion traces

chloride (Cl-): ≤0.003%

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service