Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

288446

Sigma-Aldrich

2,5-Dichlorophenyl phosphorodichloridate

97%

Synonym(s):

2,5-Dichlorophenyl dichlorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2P(O)OC6H3Cl2
CAS Number:
Molecular Weight:
279.87
Beilstein/REAXYS Number:
2118289
EC Number:
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

assay

97%

bp

141 °C/9 mmHg (lit.)

mp

36-38 °C (lit.)

SMILES string

Clc1ccc(Cl)c(OP(Cl)(Cl)=O)c1

InChI key

LYUNTJUPGQYMJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service