Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28745

Sigma-Aldrich

trans,cis,cis-1,5,9-Cyclododecatriene

≥96.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18
CAS Number:
Molecular Weight:
162.27
Beilstein/REAXYS Number:
2038580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥96.0%

refractive index

n20/D 1.513

bp

237-239 °C (lit.)

density

0.904 g/mL at 20 °C (lit.)

SMILES string

C1CC=CCC\C=C\CCC=C1

pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

190.4 °F - closed cup

flash_point_c

88 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service