Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

287245

Sigma-Aldrich

Sodium taurodeoxycholate hydrate

95%

Synonym(s):

2-([3α,12α-Dihydroxy-24-oxo-5β-cholan-24-yl]amino)ethanesulfonic acid, Taurodeoxycholic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C26H44NO6SNa · xH2O
CAS Number:
Molecular Weight:
521.69 (anhydrous basis)
Beilstein/REAXYS Number:
3901204
MDL number:
UNSPSC Code:
12161900

assay

95%

optical activity

[α]25/D +33°, c = 2.5 in H2O

mol wt

micellar avg mol wt 3100

aggregation number

6

CMC

1-4 mM (20-25°C)

SMILES string

O.[Na+].C[C@H](CCC(=O)NCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C26H45NO6S.Na.H2O/c1-16(4-9-24(30)27-12-13-34(31,32)33)20-7-8-21-19-6-5-17-14-18(28)10-11-25(17,2)22(19)15-23(29)26(20,21)3;;/h16-23,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);;1H2/q;+1;/p-1/t16-,17-,18-,19+,20-,21+,22+,23+,25+,26-;;/m1../s1

InChI key

OLPIZAYYAVQETM-GGPRKOIFSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Bile salt-related, anionic detergent used for isolation of membrane proteins including inner mitochondrial membrane proteins.

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Steil et al.
Journal of lipid research, 56(12), 2322-2336 (2015-10-16)
Shiga toxins (Stxs) are produced by enterohemorrhagic Escherichia coli (EHEC), which cause human infections with an often fatal outcome. Vero cell lines, derived from African green monkey kidney, represent the gold standard for determining the cytotoxic effects of Stxs. Despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service