Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

287113

Sigma-Aldrich

3-(2-Pyridyldithio)propionic acid N-hydroxysuccinimide ester

98%

Synonym(s):

N-Succinimidyl 3-(2-pyridyldithio)propionate, SPDP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O4S2
CAS Number:
Molecular Weight:
312.36
Beilstein/REAXYS Number:
1548137
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

98%

mp

84-86 °C (lit.)

SMILES string

O=C(CCSSC1=NC=CC=C1)ON2C(CCC2=O)=O

InChI

1S/C12H12N2O4S2/c15-10-4-5-11(16)14(10)18-12(17)6-8-19-20-9-3-1-2-7-13-9/h1-3,7H,4-6,8H2

InChI key

JWDFQMWEFLOOED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile heterofunctional cross-linking agent for proteins and enzymes.[1]

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 18, 107-107 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service