Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

286907

Sigma-Aldrich

Sodium palmitate

98%

Synonym(s):

Hexadecanoic acid sodium salt, Palmitic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COONa
CAS Number:
Molecular Weight:
278.41
Beilstein/REAXYS Number:
3575882
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

mp

283-290 °C (lit.)

SMILES string

[Na+].CCCCCCCCCCCCCCCC([O-])=O

InChI

1S/C16H32O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1

InChI key

GGXKEBACDBNFAF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service