Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

286532

Sigma-Aldrich

3-Maleimidobenzoic acid N-hydroxysuccinimide ester

97%

Synonym(s):

MBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2O6
CAS Number:
Molecular Weight:
314.25
Beilstein/REAXYS Number:
1505254
MDL number:
UNSPSC Code:
12352103

assay

97%

mp

175-177 °C (lit.)

SMILES string

O=C1CCC(N1OC(C2=CC=CC(N3C(C=CC3=O)=O)=C2)=O)=O

InChI

1S/C15H10N2O6/c18-11-4-5-12(19)16(11)10-3-1-2-9(8-10)15(22)23-17-13(20)6-7-14(17)21/h1-5,8H,6-7H2

InChI key

LLXVXPPXELIDGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.0, followed by coupling to compounds containing sulfhydryl by thioether at same pH. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

Other Notes

The aryl maleimide is less stable than alkyl maleimide linkers.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service