Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

286168

Sigma-Aldrich

2-Hydroxyestradiol

95%

Synonym(s):

1,3,5(10)-Estratriene-2,3-17β-triol, Estra-1,3,5(10)-triene-2,3,17-triol, NSC 61711

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
MDL number:
Pricing and availability is not currently available.

assay

95%

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)c(O)cc34)[C@@H]1CC[C@@H]2O

InChI

1S/C18H24O3/c1-18-7-6-11-12(14(18)4-5-17(18)21)3-2-10-8-15(19)16(20)9-13(10)11/h8-9,11-12,14,17,19-21H,2-7H2,1H3/t11-,12+,14-,17-,18-/m0/s1

InChI key

DILDHNKDVHLEQB-XSSYPUMDSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emil Jakobsen et al.
Biochemical pharmacology, 155, 92-101 (2018-06-26)
Soluble adenylate cyclase (sAC) is a non-plasma membrane-bound isoform of the adenylate cyclases signaling via the canonical second messenger, 3',5'-cyclic AMP (cAMP). sAC is involved in key physiological processes such as insulin release, sperm motility, and energy metabolism. Thus, sAC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service