285404
Cholesteryl linoleate
98%
Synonym(s):
3β-Hydroxy-5-cholestene 3-linoleate, 5-Cholesten-3β-ol 3-linoleate, Cholesteryl 9,12-octadecadienoate, Cholesteryl octadecadienoate
About This Item
Recommended Products
assay
98%
SMILES string
CCCCC\C=C/C\C=C/CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C
InChI
1S/C45H76O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-25-43(46)47-38-30-32-44(5)37(34-38)26-27-39-41-29-28-40(36(4)24-22-23-35(2)3)45(41,6)33-31-42(39)44/h11-12,14-15,26,35-36,38-42H,7-10,13,16-25,27-34H2,1-6H3/b12-11-,15-14-/t36-,38+,39+,40-,41+,42+,44+,45-/m1/s1
Inchi Key
NAACPBBQTFFYQB-LJAITQKLSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service