Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

283983

Sigma-Aldrich

4-Fluoro-7-nitrobenzofurazan

97%

Synonym(s):

NBD-F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2FN3O3
CAS Number:
Molecular Weight:
183.10
Beilstein/REAXYS Number:
1077571
MDL number:
UNSPSC Code:
12352100

assay

97%

mp

52-54 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(F)c2nonc12

InChI

1S/C6H2FN3O3/c7-3-1-2-4(10(11)12)6-5(3)8-13-9-6/h1-2H

InChI key

PGZIDERTDJHJFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful for fluorescent labeling of amino acids and amines in HPLC

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service