Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

283916

Sigma-Aldrich

N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride

98%

Synonym(s):

W-7, W7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21ClN2O2S · HCl
CAS Number:
Molecular Weight:
377.33
Beilstein/REAXYS Number:
6030174
MDL number:
Pricing and availability is not currently available.

assay

98%

SMILES string

Cl.NCCCCCCNS(=O)(=O)c1cccc2c(Cl)cccc12


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Bautista-Carbajal et al.
Virology, 501, 188-198 (2016-12-13)
Dengue virus (DENV) replicative cycle occurs in the endoplasmic reticulum where calcium ions play an important role in cell signaling. Calmodulin (CaM) is the primary sensor of intracellular Ca
Jing Yuan et al.
Nature chemical biology, 5(10), 765-771 (2009-09-08)
Studies of gene function and molecular mechanisms in Plasmodium falciparum are hampered by difficulties in characterizing and measuring phenotypic differences between individual parasites. We screened seven parasite lines for differences in responses to 1,279 bioactive chemicals. Hundreds of compounds were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service