Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

283762

Sigma-Aldrich

1,4-Diisopropyl-1-cyclohexene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22
CAS Number:
Molecular Weight:
166.30
MDL number:
UNSPSC Code:
12352103

assay

96%

SMILES string

CC(C)C1CCC(=CC1)C(C)C

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

159.8 °F - closed cup

flash_point_c

71 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service