Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

281298

Sigma-Aldrich

1,5,9,13-Tetrathiacyclohexadecane

97%

Synonym(s):

16-Ane-S4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24S4
CAS Number:
Molecular Weight:
296.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

57-58 °C (lit.)

functional group

thioether

SMILES string

C1CSCCCSCCCSCCCSC1

InChI

1S/C12H24S4/c1-5-13-7-2-9-15-11-4-12-16-10-3-8-14-6-1/h1-12H2

InChI key

LIVCAIOLNUODDU-UHFFFAOYSA-N

General description

1,5,9,13-Tetrathiacyclohexadecane is a macrocyclic tetrathioether and forms complexes with heavy metals via reaction with heavy metal salts[1]. 1,5,9,13-Tetrathiacyclohexadecane ([16]aneS4) reacts with [Ni(OH2)6][BF4]2 in MeNO2 in the presence of acetic anhydride to yield [Ni([16]aneS4)(OH2)2]2+[2].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and complexation studies of mesocyclic and macrocyclic polythioethers. 8. Synthesis and x-ray crystal structures of heavy-metal complexes of 1, 5, 9, 13-tetrathiacyclohexadecane.
Setzer WN, et al.
Inorganic Chemistry, 30(19), 3652-3656 (1991)
Nickel thioether chemistry: syntheses of nickel (II) complexes of tetra-and penta-thia macrocyclic ligands.
Blake AJ, et al.
J. Chem. Soc., Dalton Trans., 19, 2803-2808 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service