Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

275212

Sigma-Aldrich

Crotonaldehyde, predominantly trans

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCHO
CAS Number:
Molecular Weight:
70.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.41 (vs air)

vapor pressure

32 mmHg ( 20 °C)

assay

98%

autoignition temp.

320 °F

expl. lim.

19.5 %

refractive index

n20/D 1.4362 (lit.)

bp

104 °C (lit.)

mp

−76 °C (lit.)

density

0.846 g/mL at 25 °C (lit.)

SMILES string

[H]\C(C)=C(\[H])C([H])=O

InChI

1S/C4H6O/c1-2-3-4-5/h2-4H,1H3/b3-2+

InChI key

MLUCVPSAIODCQM-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

48.2 °F - closed cup

flash_point_c

9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eşref Demir et al.
Mutation research, 726(2), 98-103 (2011-07-19)
Lipid-peroxidation products are formed by the thermal treatment of foodstuffs, as well as by endogenous processes. In addition, they are also common environmental pollutants originating from many different sources. Since conflicting data exist on their possible risk for humans, we
Qian-Yuan Wu et al.
Water research, 44(12), 3625-3634 (2010-05-25)
Recent studies have reported that chlorination increased the antiestrogenic activity of wastewater, suggesting that disinfection by-products (DBPs) formed during chlorination is a potential and important source of endocrine-disruptor. However, antiestrogenic DBPs have not been identified. In this study, the antiestrogenic
Giorgia Volpi et al.
British journal of pharmacology, 163(3), 649-661 (2011-02-11)
Vascular endothelial growth factor (VEGF) is an angiogenic factor known to be elevated in the sputum of asymptomatic smokers as well as smokers with bronchitis type of chronic obstructive pulmonary disease. The aim of this study was to investigate whether
Organocatalytic asymmetric inverse-electron-demand Diels-Alder reaction of electron-deficient dienes and crotonaldehyde.
Jun-Long Li et al.
Angewandte Chemie (International ed. in English), 49(36), 6418-6420 (2010-07-31)
Thomas W Kensler et al.
Carcinogenesis, 33(1), 101-107 (2011-11-03)
Epidemiological evidence has suggested that consumption of a diet rich in cruciferous vegetables reduces the risk of several types of cancers and chronic degenerative diseases. In particular, broccoli sprouts are a convenient and rich source of the glucosinolate, glucoraphanin, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service