Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

273198

Sigma-Aldrich

2-Chloro-4-methylphenol

97%

Synonym(s):

2-Chloro-p-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)OH
CAS Number:
Molecular Weight:
142.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.553 (lit.)

bp

195-196 °C (lit.)

density

1.211 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(O)c(Cl)c1

InChI

1S/C7H7ClO/c1-5-2-3-7(9)6(8)4-5/h2-4,9H,1H3

InChI key

AQJFATAFTQCRGC-UHFFFAOYSA-N

General description

The rate of photocatalytic degradation of 2-chloro-4-methylphenol was studied[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

186.8 °F - closed cup

flash_point_c

86 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of photocatalytic degradation of phenols with multiple substituent groups.
Journal of Photochemistry and Photobiology A: Chemistry, 179(3), 256-262 (2006)
Liancheng Fang et al.
Journal of hazardous materials, 388, 121787-121787 (2019-12-11)
Dihalophenols such as dichlorophenols (DCPs) are important industrial chemical intermediates, but also persistent pollutants in the environment. Oxidative dehalogenation by microbes is an efficient biological method to degrade halophenols, but the mechanism is unclear yet. Cupriavidus nantongensis X1T was a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service