271497
Dihexadecyl phosphate
99%
Synonym(s):
Dicetyl phosphate
About This Item
assay
99%
mp
74-75 °C (lit.)
SMILES string
OP(OCCCCCCCCCCCCCCCC)(OCCCCCCCCCCCCCCCC)=O
InChI
1S/C32H67O4P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-35-37(33,34)36-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-32H2,1-2H3,(H,33,34)
InChI key
RNPXCFINMKSQPQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service