Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

270431

Supelco

Dimethyl sulfoxide

HPLC grade, 99.9%

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein/REAXYS Number:
506008
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

grade

HPLC grade

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

assay

99.9%

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

impurities

<0.050% water

evapn. residue

<0.0005%

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)
189 °C

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

λ

H2O reference

UV absorption

λ: 270 nm Amax: 1.00
λ: 280 nm Amax: 0.30
λ: 295 nm Amax: 0.10
λ: 350-400 nm Amax: 0.01

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

Inchi Key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia C Faria et al.
Cancer research, 75(1), 134-146 (2014-11-14)
Medulloblastoma is the most common malignant pediatric brain tumor, with metastases present at diagnosis conferring a poor prognosis. Mechanisms of dissemination are poorly understood and metastatic lesions are genetically divergent from the matched primary tumor. Effective and less toxic therapies
Rory W Blackler et al.
British journal of pharmacology, 172(4), 992-1004 (2014-10-10)
Hydrogen sulphide is an important mediator of gastrointestinal mucosal defence. The use of non-steroidal anti-inflammatory drugs (NSAIDs) is significantly limited by their toxicity in the gastrointestinal tract. Particularly concerning is the lack of effective preventative or curative treatments for NSAID-induced
Hongjie An et al.
Advances in colloid and interface science, 222, 9-17 (2014-08-17)
The observation by Atomic Force Microscopy of a range of nanophases on hydrophobic surfaces poses some challenging questions, not only related to the stability of these objects but also regarding their wetting properties. Spherical capped nanobubbles are observed to exhibit
Mulmudi Hemant Kumar et al.
Advanced materials (Deerfield Beach, Fla.), 26(41), 7122-7127 (2014-09-13)
Lead free perovskite solar cells based on a CsSnI3 light absorber with a spectral response from 950 nm is demonstrated. The high photocurrents noted in the system are a consequence of SnF2 addition which reduces defect concentrations and hence the
Björn Zörner et al.
Brain : a journal of neurology, 137(Pt 6), 1716-1732 (2014-04-17)
Anatomical plasticity such as fibre growth and the formation of new connections in the cortex and spinal cord is one known mechanism mediating functional recovery after damage to the central nervous system. Little is known about anatomical plasticity in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service