About This Item
Recommended Products
grade
HPLC grade
vapor density
2.48 (vs air)
vapor pressure
26.85 psi ( 55 °C)
8.43 psi ( 20 °C)
assay
≥99%
autoignition temp.
500 °F
purified by
glass distillation
expl. lim.
~8.3 %
impurities
<0.010% water
evapn. residue
<0.0003%
refractive index
n20/D 1.358 (lit.)
bp
35-36 °C (lit.)
mp
−130 °C (lit.)
density
0.626 g/mL at 25 °C (lit.)
λ
H2O reference
UV absorption
λ: 195 nm Amax: 1.00
λ: 200 nm Amax: 0.60
λ: 210 nm Amax: 0.30
λ: 220 nm Amax: 0.06
λ: 245-400 nm Amax: 0.01
SMILES string
CCCCC
InChI
1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3
InChI key
OFBQJSOFQDEBGM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
-40.0 °F
flash_point_c
-40 °C
ppe
Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service