270075
1-(α,α,α-Trifluoro-m-tolyl)piperazine hydrochloride
99%
Synonym(s):
N-[3-(Trifluoromethyl)phenyl]piperazine hydrochloride, TFMPP hydrochloride
About This Item
assay
99%
drug control
regulated under CDSA - not available from Sigma-Aldrich Canada
mp
239-241 °C (lit.)
SMILES string
Cl.FC(F)(F)c1cccc(c1)N2CCNCC2
InChI
1S/C11H13F3N2.ClH/c12-11(13,14)9-2-1-3-10(8-9)16-6-4-15-5-7-16;/h1-3,8,15H,4-7H2;1H
InChI key
DGNLGWJZZZOYPT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service