Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

269808

Sigma-Aldrich

N,N-Dimethylformamide-d7

99.5 atom % D

Synonym(s):

DMF-d7, Heptadeutero-N,N-dimethylformamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCON(CD3)2
CAS Number:
Molecular Weight:
80.14
Beilstein/REAXYS Number:
1908468
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.5 atom % D

Quality Level

assay

99% (CP)

packaging

pk of 10 × 1 mL

refractive index

n20/D 1.428 (lit.)

bp

153 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

mass shift

M+7

SMILES string

[2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i1D3,2D3,3D

InChI key

ZMXDDKWLCZADIW-YYWVXINBSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

135.5 °F - closed cup

flash_point_c

57.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Małgorzata Fabijańska et al.
Dalton transactions (Cambridge, England : 2003), 44(3), 938-947 (2014-08-12)
This paper describes the synthesis of trans-bis-(3-aminoflavone)dichloridoplatinum(ii) (trans-Pt(3-af)2Cl2; TCAP) for use as a potential anticancer compound, and the evaluation of its structure by elemental and spectral analyses, and X-ray crystallography. The complex demonstrated a significant cytotoxic effect against human and
Seung Bin Baek et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(46), 14156-14161 (2015-11-19)
Crystallographic observation of adsorbed gas molecules is a highly difficult task due to their rapid motion. Here, we report the in situ single-crystal and synchrotron powder X-ray observations of reversible CO2 sorption processes in an apparently nonporous organic crystal under
Sean E Doris et al.
Journal of the American Chemical Society, 136(44), 15702-15710 (2014-10-11)
Cationic naked nanocrystals (NCs) are useful building units for assembling hierarchical mesostructured materials. Until now, their preparation required strongly electrophilic reagents that irreversibly sever bonds between native organic ligands and the NC surface. Colloidal instabilities can occur during ligand stripping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service