Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

269778

Sigma-Aldrich

Trifluoroacetic acid-d

99.5 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOD
CAS Number:
Molecular Weight:
115.03
Beilstein/REAXYS Number:
1768662
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
Product 269778 is not currently sold in your country. Contact Technical Service

isotopic purity

99.5 atom % D

Quality Level

form

liquid

mol wt

115.02 by atom % calculation

packaging

pk of 10 × 1 mL

technique(s)

NMR: suitable

impurities

≤0.500% water
water

refractive index

n20/D 1.3 (lit.)

bp

75 °C (lit.)

mp

-15 °C (lit.)

density

1.493 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OC(=O)C(F)(F)F

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)/i/hD

InChI key

DTQVDTLACAAQTR-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti stereochemistry in protodesilylation of an optically active allylsilane with trifluoroacetic acid-D.
Hayashi T, et al.
Tetrahedron Letters, 23(44), 4605-4606 (1982)
The Vapor Phase Dissociation of Some Carboxylic Acids. III. 1 Trifluoroacetic Acid and Trifluoroacetic Acid-d2, 3.
Taylor MD and Templeman MB.
Journal of the American Chemical Society, 78(13), 2950-2953 (1956)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service