266604
Antimony
beads, 1-2 mm, 99.999% trace metals basis
Synonym(s):
Antimony black, Antimony element
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Sb
CAS Number:
Molecular Weight:
121.76
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay
99.999% trace metals basis
form
beads
resistance
41.7 μΩ-cm
particle size
1-2 mm
bp
1635 °C (lit.)
mp
630 °C (lit.)
density
6.69 g/mL at 25 °C (lit.)
SMILES string
[Sb]
InChI
1S/Sb
InChI key
WATWJIUSRGPENY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
application
Applied in the solid state synthesis of a Zintl phase, SrSn3Sb4, with an anionic channel framework of 30-membered rings.
replaced by
Product No.
Description
Pricing
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jaime Soto et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 56(9), 1255-1260 (2013-02-08)
Cutaneous leishmaniasis is an ultimately self-curing disease for which systemic therapy with pentavalent antimony (Sb) is effective but with side effects. We evaluated 2 local treatments, intralesional (IL) Sb and cryotherapy, for single lesions due to Bolivian Leishmania (v.) braziliensis
Jian Chen et al.
Chemosphere, 90(6), 1925-1932 (2012-11-15)
Coal is one of the major energy resources in China, with nearly half of produced Chinese coal used for power and heat generation. The large use of coal for power and heat generation in China may result in significant atmospheric
Joseph N Jarvis et al.
Current opinion in infectious diseases, 26(1), 1-9 (2012-12-12)
HIV infection profoundly impairs the immune mechanisms needed to control and clear Leishmania infection, and outcomes in patients with HIV-associated visceral leishmaniasis are poor. This review summarizes recent work describing the epidemiology, presentation and outcomes of HIV-associated visceral leishmaniasis and
Konstantinos C Makris et al.
Journal of exposure science & environmental epidemiology, 23(2), 120-127 (2012-11-29)
Environmental factors, such as storage time, frequency of bottle reuse and temperature, have been shown to facilitate antimony (Sb) leaching from water- and food-packaging materials. The globally escalating consumption of water packaged in Sb-containing bottles, such as that of polyethylene
Anette Stauch et al.
PLoS neglected tropical diseases, 6(12), e1973-e1973 (2013-01-04)
Pentavalent antimonials have been the mainstay of antileishmanial therapy for decades, but increasing failure rates under antimonial treatment have challenged further use of these drugs in the Indian subcontinent. Experimental evidence has suggested that parasites which are resistant against antimonials
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service