Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

265357

Sigma-Aldrich

Cadmium

ReagentPlus®, 99.5% trace metals basis, powder, −325 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
Beilstein/REAXYS Number:
8137359
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

product line

ReagentPlus®

assay

99.5% trace metals basis

form

powder

resistivity

7.27 μΩ-cm, 22°C

particle size

−325 mesh

bp

765 °C (lit.)

mp

320.9 °C (lit.)

density

8.65 g/mL at 25 °C (lit.)

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiping Chen et al.
Environmental pollution (Barking, Essex : 1987), 178, 294-299 (2013-04-18)
Safety of agricultural irrigation with reclaimed water is of great concern as some potential hazardous compounds like heavy metals may be accumulated in soils over time. Impacts of long-term reclaimed water on soil Cd pollution were evaluated based on the
Shigeo Hasegawa et al.
Molecular pain, 5, 22-22 (2009-05-05)
Peripheral nerve injury leads to a persistent neuropathic pain state in which innocuous stimulation elicits pain behavior (tactile allodynia), but the underlying mechanisms have remained largely unknown. We have previously shown that spinal nerve injury induces the activation of cytosolic
F Tomei et al.
Annali di igiene : medicina preventiva e di comunita, 25(4), 353-363 (2013-05-25)
The exposure to Cd has been linked to many disease and liver is the major target organ of Cd toxicity following acute or chronic exposure.No study has ever evaluated the association between liver function tests and exposure to Cd present
Nivedita P Khairnar et al.
Journal of bacteriology, 195(12), 2880-2886 (2013-04-23)
Deinococcus radiodurans R1 exposed to a lethal dose of cadmium shows differential expression of a large number of genes, including frnE (drfrnE) and some of those involved in DNA repair and oxidative stress tolerance. The drfrnE::nptII mutant of D. radiodurans
Gang Zhang et al.
Journal of environmental sciences (China), 25(1), 231-235 (2013-04-17)
The emission characteristics of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/Fs) and heavy metals were evaluated during co-combustion of sewage sludge with coal from a circulating fluidized bed incinerator. The stack gas, slag and fly ash samples were sampled and analyzed. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service