Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

264830

Sigma-Aldrich

Antimony

beads, −4-20 mesh, 99.9998% trace metals basis

Synonym(s):

Antimony black, Antimony element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sb
CAS Number:
Molecular Weight:
121.76
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.9998% trace metals basis

form

beads

resistance

41.7 μΩ-cm

particle size

−4-20 mesh

bp

1635 °C (lit.)

mp

630 °C (lit.)

density

6.69 g/mL at 25 °C (lit.)

SMILES string

[Sb]

InChI

1S/Sb

InChI key

WATWJIUSRGPENY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Applied in the solid state synthesis of a Zintl phase, SrSn3Sb4, with an anionic channel framework of 30-membered rings.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daoyong Zhang et al.
Bioresource technology, 128, 711-715 (2012-12-19)
Extracellular polymeric substances (EPS) existed ubiquitously in biological systems affect the mobility and availability of heavy metals in the environments. The adsorption-desorption behaviors of Hg(II) and Sb(V) on EPS were investigated. The sorption rates follow Sb(V) > Hg(II), and the
Budhaditya Mukherjee et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(7), E575-E582 (2013-01-24)
The molecular mechanism of antimony-resistant Leishmania donovani (Sb(R)LD)-driven up-regulation of IL-10 and multidrug-resistant protein 1 (MDR1) in infected macrophages (Ms) has been investigated. This study showed that both promastigote and amastigote forms of Sb(R)LD, but not the antimony-sensitive form of
Konstantinos C Makris et al.
Journal of exposure science & environmental epidemiology, 23(2), 120-127 (2012-11-29)
Environmental factors, such as storage time, frequency of bottle reuse and temperature, have been shown to facilitate antimony (Sb) leaching from water- and food-packaging materials. The globally escalating consumption of water packaged in Sb-containing bottles, such as that of polyethylene
Jaime Soto et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 56(9), 1255-1260 (2013-02-08)
Cutaneous leishmaniasis is an ultimately self-curing disease for which systemic therapy with pentavalent antimony (Sb) is effective but with side effects. We evaluated 2 local treatments, intralesional (IL) Sb and cryotherapy, for single lesions due to Bolivian Leishmania (v.) braziliensis
Joseph N Jarvis et al.
Current opinion in infectious diseases, 26(1), 1-9 (2012-12-12)
HIV infection profoundly impairs the immune mechanisms needed to control and clear Leishmania infection, and outcomes in patients with HIV-associated visceral leishmaniasis are poor. This review summarizes recent work describing the epidemiology, presentation and outcomes of HIV-associated visceral leishmaniasis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service