264121
Boron trifluoride-methanol solution
in excess methanol, BF3 12 wt. %
About This Item
Recommended Products
vapor pressure
19.5 mmHg ( 20 °C)
autoignition temp.
788 °F
composition
BF3, 12 wt. %
concentration
in excess methanol
refractive index
n20/D 1.328
density
0.868 g/mL at 25 °C
SMILES string
CO.FB(F)F
InChI
1S/CH4O.BF3/c1-2;2-1(3)4/h2H,1H3;
InChI key
JBXYCUKPDAAYAS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service