262765
Niobium
wire, wire diam. 1.0 mm, 99.8% trace metals basis
Synonym(s):
Columbium
About This Item
assay
99.8% trace metals basis
form
wire
resistivity
13-16 μΩ-cm, 20°C
wire diam.
1.0 mm
bp
4742 °C (lit.)
mp
2468 °C (lit.)
density
8.57 g/mL at 25 °C (lit.)
SMILES string
[Nb]
InChI
1S/Nb
InChI key
GUCVJGMIXFAOAE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Quantity
Storage Class
13 - Non Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service