Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

261246

Sigma-Aldrich

Scandium

dendritic pieces, purified by distillation, 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sc
CAS Number:
Molecular Weight:
44.96
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.9% trace rare earth metals basis

form

dendritic pieces

purified by

distillation

resistivity

50.5 μΩ-cm, 0°C

bp

2836 °C (lit.)

mp

1540 °C (lit.)

density

2.99 g/mL at 25 °C (lit.)

SMILES string

[Sc]

InChI

1S/Sc

Inchi Key

SIXSYDAISGFNSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in solid state synthesis of unusual clusters, Sc19Br28Z4, (Z=Mn, Fe, Ru or Os). These clusters are of interest for their structure and magnetic properties.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Tran et al.
Organic letters, 14(23), 6100-6103 (2012-11-23)
Treatment of ynol ether-tethered dialkyl acetals with catalytic quantities of scandium triflate in CH(3)CN gives rise to five-, six-, and seven-membered alkoxycycloalkene carboxylates in good to excellent yields. Tri- and tetrasubstituted carbocyclic and heterocyclic alkenes may be formed by this
Marek Pruszyński et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 70(6), 974-979 (2012-04-03)
The positron-emitting radionuclide (44)Sc with a half-life of 3.97 h and a β(+) branching of 94.3% is of potential interest for clinical PET. As so far it is available from a (44)Ti/(44)Sc generator in Mainz, where long-lived (44)Ti decays to
Nadine V Hanhan et al.
Organic letters, 14(9), 2218-2221 (2012-04-18)
The scandium(III)-catalyzed enantioselective Hosomi-Sakurai allylation of isatins with various substituted allylic silanes is described. A catalyst loading as low as 0.05 mol % is utilized at room temperature to afford the 3-allyl-3-hydroxy-2-oxindoles in excellent yields and enantioselectivity up to 99%
Yunfei Cai et al.
Organic letters, 14(11), 2726-2729 (2012-05-17)
Catalytic asymmetric sulfenylation of unprotected 3-substituted oxindoles has been developed via cooperative catalysis of a chiral N,N'-dioxide-Sc(OTf)(3) complex and a Brønsted base. Utilizing readily available N-(phenylthio)phthalimide as the sulfur source, a wide range of optically active 3-phenylthiooxindoles were obtained in
Jacob S Kanady et al.
Journal of the American Chemical Society, 135(3), 1073-1082 (2012-12-18)
The oxygen-evolving complex (OEC) of photosystem II contains a Mn(4)CaO(n) catalytic site, in which reactivity of bridging oxidos is fundamental to OEC function. We synthesized structurally relevant cuboidal Mn(3)MO(n) complexes (M = Mn, Ca, Sc; n = 3,4) to enable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service