Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

260940

Sigma-Aldrich

Thianthrene

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8S2
CAS Number:
Molecular Weight:
216.32
Beilstein/REAXYS Number:
83046
EC Number:
MDL number:
UNSPSC Code:
12352100

assay

≥99%

bp

364-366 °C (lit.)

mp

151-155 °C (lit.)

SMILES string

S1c2ccccc2Sc3ccccc13

InChI

1S/C12H8S2/c1-2-6-10-9(5-1)13-11-7-3-4-8-12(11)14-10/h1-8H

InChI key

GVIJJXMXTUZIOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Experiences with thianthrol salve in dermatology].
C SEROWY
Zeitschrift fur Haut- und Geschlechtskrankheiten, 10(9), 370-376 (1951-05-01)
L L Gershbein
Research communications in chemical pathology and pharmacology, 53(1), 125-128 (1986-07-01)
Male rats were fed a diet supplemented with 0.12% thianthrene or control chow for a period of 10 days after which they were partially hepatectomized, two-thirds of the organ being removed and continued on the diet or switched to the
S C Mitchell et al.
Xenobiotica; the fate of foreign compounds in biological systems, 28(1), 93-101 (1998-03-11)
1. Radiolabelled thianthrene was administered by gavage (200 mg/kg body weight) to the adult female Wistar rat following an overnight fast. 2. Faeces was the major route of excretion of radioactivity (62%) with substantial amounts still being voided during the
Formation of thianthrene by a free radical mechanism.
D A SHIRLEY et al.
Science (New York, N.Y.), 113(2930), 208-210 (1951-02-23)
Patrick Frank et al.
Inorganic chemistry, 45(24), 9864-9876 (2006-11-23)
Sulfur K-edge X-ray absorption spectroscopy (XAS) was used to characterize the approximately 0.1% sulfur found both in native reticulated vitreous carbon (RVC) foam and in RVC oxidatively modified using 0.2 M KMnO4 in 2 M H2SO4. Sulfur valences and functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service