Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

258458

Sigma-Aldrich

Bis(triphenylphosphoranylidene)ammonium borohydride

97%

Synonym(s):

Bis(triphenylphosphine)iminium borohydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H34BNP2
CAS Number:
Molecular Weight:
553.42
MDL number:
UNSPSC Code:
12352103

assay

97%

SMILES string

[BH4-].c1ccc(cc1)P(=[N+]=P(c2ccccc2)(c3ccccc3)c4ccccc4)(c5ccccc5)c6ccccc6

Pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service