Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257036

Sigma-Aldrich

Isopropylmagnesium chloride solution

2.0 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHMgCl
CAS Number:
Molecular Weight:
102.85
Beilstein/REAXYS Number:
4124221
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

concentration

2.0 M in THF

density

0.975 g/mL at 25 °C

SMILES string

CC(C)[Mg]Cl

InChI

1S/C3H7.ClH.Mg/c1-3-2;;/h3H,1-2H3;1H;/q;;+1/p-1

InChI key

GPIBKUJXKCEZOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service