Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

256927

Sigma-Aldrich

Ethylaluminum dichloride solution

1.0 M in hexanes

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2AlCl2
CAS Number:
Molecular Weight:
126.95
Beilstein/REAXYS Number:
4123357
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

vapor pressure

5 mmHg ( 60 °C)

form

liquid

concentration

1.0 M in hexanes

bp

68-70 °C

density

0.729 g/mL at 25 °C

SMILES string

CC[Al](Cl)Cl

InChI

1S/C2H5.Al.2ClH/c1-2;;;/h1H2,2H3;;2*1H/q;+2;;/p-2

InChI key

UAIZDWNSWGTKFZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3 - Water-react 1

target_organs

Nervous system, Respiratory system

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barry B Snider et al.
The Journal of organic chemistry, 68(16), 6451-6454 (2003-08-05)
EtAlCl(2)-induced cyclization of chiral gamma,delta-unsaturated ketones 11c and 17b takes place mainly from the expected face. The selectivity is modest for 11c (60:40) in which the large substituent is a primary alkyl group and the medium substituent is a methyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service