Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

256897

Sigma-Aldrich

Dimethylaluminum chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2AlCl
CAS Number:
Molecular Weight:
92.50
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

79 mmHg ( 60 °C)

assay

97%

bp

126-127 °C (lit.)

mp

−21 °C (lit.)

density

0.996 g/mL at 25 °C (lit.)

SMILES string

C[Al](C)Cl

InChI

1S/2CH3.Al.ClH/h2*1H3;;1H/q;;+1;/p-1

InChI key

JGHYBJVUQGTEEB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 1

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service