256692
Cyclohexylmagnesium chloride solution
2.0 M in diethyl ether
About This Item
Recommended Products
reaction suitability
reaction type: Grignard Reaction
concentration
2.0 M in diethyl ether
density
0.871 g/mL at 25 °C
SMILES string
Cl[Mg]C1CCCCC1
InChI
1S/C6H11.ClH.Mg/c1-2-4-6-5-3-1;;/h1H,2-6H2;1H;/q;;+1/p-1
InChI key
WMJMABVHDMRMJA-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Packaging
Compatible with the following:
- Aldrich® Sure/Pac™ station for liquefied gases Z566446
- PTFE Sealing tape Z104388 or Z221880
- Straight septum-inlet adapter Z118141 with septa Z565687 or Z565695.
Legal Information
also commonly purchased with this product
recommended
septum inlet adapter
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
-40.0 °F
flash_point_c
-40 °C
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service