Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

255122

Sigma-Aldrich

1,2-Benzodiphenylene sulfide

99%

Synonym(s):

Benzo[b]naphtho[2,1-d]thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10S
CAS Number:
Molecular Weight:
234.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

solid

mp

188-190 °C (lit.)

solubility

chloroform: soluble 2.5%, clear to very slightly hazy, colorless to faint yellow or tan

SMILES string

c1ccc2c(c1)ccc3c4ccccc4sc23

InChI

1S/C16H10S/c1-2-6-12-11(5-1)9-10-14-13-7-3-4-8-15(13)17-16(12)14/h1-10H

InChI key

YEUHHUCOSQOCIX-UHFFFAOYSA-N

General description

1,2-benzodiphenylene sulfide is a rare constituent of coal tar. Photodeoxygenation of 1,2-benzodiphenylene sulfoxide, generates an intermediate capable of oxidizing the solvent benzene to phenol[1]. Liquid-phase photolysis of 1,2-benzodiphenylene sulfide to generate atomic oxygen [O(3P)] or an equivalent active oxygen species has been reported[2].

Application

1,2-Benzodiphenylene sulfide has been used in direct screening of sediment samples for polycyclic aromatic hydrocarbons by flow manifold coupled on-line to a fluorimetric detector[3].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct automatic screening and individual determination of polycyclic aromatic hydrocarbons using supercritical fluid extraction coupled on-line with liquid chromatography and fluorimetric detection.
Zougagh M, et al.
Analytica Chimica Acta, 524(1), 279-285 (2004)
Keith B Thomas et al.
The Journal of organic chemistry, 68(5), 1886-1891 (2003-03-01)
A liquid-phase photolysis of 1,2-benzodiphenylene sulfoxide, 1, and dibenzothiophene sulfoxide, 2, was used to generate atomic oxygen [O(3P)] or an equivalent active oxygen species. The reaction for sulfoxide photodeoxygenation was similar to a microwave discharge method for generating O(3P) atoms
E Lucien et al.
The Journal of organic chemistry, 66(13), 4576-4579 (2001-06-26)
We report that the photodeoxygenation of 1,2-benzodiphenylene sulfoxide, 1, generates an intermediate capable of oxidizing the solvent benzene to phenol. The reactivity of the intermediate was probed with various substrates (2-methylbutane, chloride ion, and para-substituted aryl sulfides). The intermediate produced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service