Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

253588

Sigma-Aldrich

Methyl 12-DOXYL-stearate, free radical

Synonym(s):

2-Hexyl-2-(11-methoxy-11-oxoundecyl)-4,4-dimethyl-3-oxazolidinyloxy, 2-Hexyl-2-(11-methoxy-11-oxoundecyl)-4,4-dimethyl-3-oxazolidinyloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H45NO4
CAS Number:
Molecular Weight:
399.61
MDL number:
UNSPSC Code:
12352200

SMILES string

CCCCCCC1(CCCCCCCCCCC(=O)OC)OCC(C)(C)N1[O]

Features and Benefits

Spin label.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mobility of apolar lipids of reconstituted low density lipoprotein as monitored by electron spin resonance spectroscopy.
M Krieger et al.
The Journal of biological chemistry, 255(8), 3330-3333 (1980-04-25)
E Grzelińska et al.
International journal of radiation biology and related studies in physics, chemistry, and medicine, 42(1), 45-55 (1982-07-01)
Spin-label studies of the effects of hyperthermia on the erythrocyte membrane revealed a decrease in the fluidity of lipids and changes in the state of membrane proteins. The rate of haemolysis in iso-osmotic glycerol solution was increased. Changes of most
M Derby et al.
Cytobios, 102(400), 115-118 (2000-07-08)
Rabbit erythrocytes in methanolic phosphate medium were used to bioassay the activity of authentic samples of methyl stearate and methyl palmitate (in 10% methanol:90% water, v/v), which had been identified as apparent oceanic naturally occurring cytolins (APONIN-3 and -4) produced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service