Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

251763

Sigma-Aldrich

trans-2-Heptenal

90%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CHCHO
CAS Number:
Molecular Weight:
112.17
Beilstein/REAXYS Number:
1700822
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

vapor density

>1 (vs air)

assay

90%

refractive index

n20/D 1.450 (lit.)

bp

90-91 °C/50 mmHg (lit.)

density

0.857 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)\C([H])=C(/[H])CCCC

InChI

1S/C7H12O/c1-2-3-4-5-6-7-8/h5-7H,2-4H2,1H3/b6-5+

InChI key

NDFKTBCGKNOHPJ-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service