251399
Trichloroacetic acid
ACS reagent, ≥99%
Synonym(s):
TCA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Cl3CCOOH
CAS Number:
Molecular Weight:
163.39
Beilstein/REAXYS Number:
970119
EC Number:
MDL number:
UNSPSC Code:
12352100
Recommended Products
grade
ACS reagent
vapor density
<1 (vs air)
<1 (vs air)
vapor pressure
1 mmHg ( 51 °C)
assay
≥99%
ign. residue
≤0.03%
refractive index
n20/D 1.62 (lit.)
bp
196 °C (lit.)
mp
54-58 °C (lit.)
density
1.62 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.02%
cation traces
Fe: ≤0.001%
heavy metals: ≤0.002%
SMILES string
OC(=O)C(Cl)(Cl)Cl
InChI
1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)
InChI key
YNJBWRMUSHSURL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service