Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

251097

Sigma-Aldrich

1-(4-Iodophenyl)-5-(4-nitrophenyl)-3-phenylformazan

Synonym(s):

INT Formazan, Iodonitrotetrazolium formazan, Iodonitrotetrazolium violet formazan, Iodonitrotetrazoliumviolet-formazan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4N=NC(C6H5)=NNHC6H4NO2
CAS Number:
Molecular Weight:
471.25
Beilstein/REAXYS Number:
1829738
EC Number:
MDL number:
UNSPSC Code:
12171500

mp

187 °C (dec.) (lit.)

SMILES string

[O-][N+](=O)c1ccc(NN=C(N=Nc2ccc(I)cc2)c3ccccc3)cc1

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service