Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

250562

Sigma-Aldrich

2-Methoxy-3-methylpyrazine

technical grade, 75%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O
CAS Number:
Molecular Weight:
124.14
MDL number:
Pricing and availability is not currently available.

grade

technical grade

assay

75%

refractive index

n20/D 1.507 (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

COc1nccnc1C

InChI

1S/C6H8N2O/c1-5-6(9-2)8-4-3-7-5/h3-4H,1-2H3

InChI key

VKJIAEQRKBQLLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains 2,6-isomers
remainder 2,5-isomers

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service