Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

250139

Sigma-Aldrich

2,3-Dichloropropionyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2ClCHClCOCl
CAS Number:
Molecular Weight:
161.41
Beilstein/REAXYS Number:
506750
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.477 (lit.)

bp

53-54 °C/17 mmHg (lit.)

density

1.476 g/mL at 25 °C (lit.)

SMILES string

ClCC(Cl)C(Cl)=O

InChI

1S/C3H3Cl3O/c4-1-2(5)3(6)7/h2H,1H2

InChI key

JQELECXPPAOSTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service