Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

249068

Sigma-Aldrich

cis-11-Tetradecen-1-yl acetate

95%

Synonym(s):

(11Z)-11-Tetradecen-1-ol acetate, (Z)-11-TDA, Z11-14: Acetate, Z-11-TDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30O2
CAS Number:
Molecular Weight:
254.41
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

95%

SMILES string

CC\C=C/CCCCCCCCCCOC(C)=O

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

141.8 °F - closed cup

flash_point_c

61 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongping Huang et al.
Journal of chemical ecology, 28(3), 533-539 (2002-04-12)
Individual analysis of the female sex pheromone of the adzuki bean borer, Ostrinia scapulalis, has shown that the sex pheromone of this species comprised (E)-11-tetradecenyl acetate (E11-14:OAc) and (Z)-11-tetradecenyl acetate (Z11-14:OAc) at variable blend ratios. The pheromone blend could be
Ray L Hong et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(22), 7779-7784 (2008-05-30)
The geographical mosaic theory of coevolution predicts that different local species interactions will shape population traits, but little is known about the molecular factors involved in mediating the specificity of these interactions. Pristionchus nematodes associate with different scarab beetles around
Chang Yeol Yang et al.
Journal of chemical ecology, 35(3), 342-348 (2009-02-18)
We tested differences in female pheromone production and male response in three species of the genus Adoxophyes in Korea. Females of all three species produced mixtures of (Z)-9-tetradecenyl acetate (Z9-14:OAc) and (Z)-11-tetradecenyl acetate (Z11-14:OAc) as major components but in quite
S P Foster
Archives of insect biochemistry and physiology, 43(3), 125-136 (2000-02-24)
Pheromone titer in moths is a product of three processes occurring in or at the surface of the pheromone gland: biosynthesis, release, and intraglandular degradation, of pheromone. Changes in titers of sex pheromone, the fatty acyl pheromone analog (FAPA), and
J Zhu et al.
Genetics, 144(2), 757-766 (1996-10-01)
The major difference in pheromone production between the so-called E and Z strains of the European corn borer Ostrinia nubilalis is controlled by two alleles at a single autosomal locus. E-strain females produce an (E)-11-tetradecenyl acetate pheromone with 1-3% of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service