Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

249033

Sigma-Aldrich

cis-9-Tetradecenol

96%

Synonym(s):

(Z)-9-TDOL, Myristoleyl alcohol, Z-9-TDOL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH(CH2)8OH
CAS Number:
Molecular Weight:
212.37
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.457 (lit.)

SMILES string

[H]\C(CCCC)=C(/[H])CCCCCCCCO

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pheromone for Homoeosoma electellum

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service