Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24855

Sigma-Aldrich

4-Chloro-N-(2-hydroxyethyl)-2-nitroaniline

≥98.0%

Synonym(s):

2-(4-Chloro-2-nitroanilino)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9ClN2O3
CAS Number:
Molecular Weight:
216.62
Beilstein/REAXYS Number:
2213635
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98.0%

mp

104-108 °C

SMILES string

OCCNc1ccc(Cl)cc1[N+]([O-])=O

InChI

1S/C8H9ClN2O3/c9-6-1-2-7(10-3-4-12)8(5-6)11(13)14/h1-2,5,10,12H,3-4H2

InChI key

LGGKGPQFSCBUOR-UHFFFAOYSA-N

Application

4-Chloro-N-(2-hydroxyethyl)-2-nitroaniline may be used in chemical synthesis.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service