Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

248134

Sigma-Aldrich

3-Chloro-7-hydroxy-4-methylcoumarin

96%

Synonym(s):

3-Chloro-4-methylumbelliferone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7ClO3
CAS Number:
Molecular Weight:
210.61
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

assay

96%

mp

240-244 °C (lit.)

SMILES string

CC1=C(Cl)C(=O)Oc2cc(O)ccc12

InChI

1S/C10H7ClO3/c1-5-7-3-2-6(12)4-8(7)14-10(13)9(5)11/h2-4,12H,1H3

InChI key

ODZHLDRQCZXQFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J K Malik et al.
Archives of toxicology, 48(1), 51-59 (1981-08-01)
The biotransformation and disposition of 3-chloro-4-methyl-(4-14C)-7-hydroxycoumarin [(14C) chlorferron] were investigated in rats after single oral administration. Following administration of (14C) chlorferron at 0.5 and 20 mg/kg body weight to male rats, greater than 90% of the given dose was eliminated
J K Malik et al.
Advances in experimental medicine and biology, 136 Pt A, 839-846 (1981-01-01)
The distribution, excretion and biotransformation of 3-chloro-4-methyl-(4-14C)-7-hydroxycoumarin [(14C) chlorferron] were studied in the rats after single oral administration. Male and female Wistar rats were given single oral doses of 0.5 and 20.0 mg/kg body weight of (14C) chlorferron and routes
B China Raju et al.
Bioorganic & medicinal chemistry, 18(1), 358-365 (2009-11-26)
Series of 3,4- and 3,6-disubstituted chromenones including new chromenone derivatives were synthesized applying various synthetic strategies including Pechmann condensation, Knoevenagel condensation, Reimer-Tiemann reaction and Suzuki coupling in very good yields. Synthesized compounds (4a-z) were screened for in vitro alpha-glucosidase inhibitory
Jiyeon Ha et al.
Bioresource technology, 98(10), 1916-1923 (2006-09-22)
Chlorferon and diethylthiophosphate (DETP) are the hydrolysis products of coumaphos, an organophosphate pesticide. In this research, two consortia of bacterial cultures, one responsible for degrading chlorferon and the other for degrading DETP, were selectively enriched from waste cattle dip solution.
V Simeon-Rudolf et al.
Chemico-biological interactions, 119-120, 119-128 (1999-07-27)
Inhibition of recombinant mouse wild type AChE (EC 3.1.1.7) and BChE (EC 3.1.1.8), and AChE peripheral site-directed mutants and human serum BChE variants by 4,4'-bipyridine (4,4'-BP) and the coumarin derivative 3-chloro-7-hydroxy-4-methylcoumarin (CHMC) was studied. The enzyme activity was measured with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service