Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

246581

Sigma-Aldrich

Scopoletin

95%

Synonym(s):

6-Methoxyumbelliferone, 6-Methylesculetin, 7-Hydroxy-6-methoxy-2H-1-benzopyran-2-one, 7-Hydroxy-6-methoxycoumarin, Buxuletin, Chrysatropic acid, Escopoletin, Esculetin-6-methyl ether, Gelseminic acid, Murrayetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
Beilstein/REAXYS Number:
156296
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

95%

mp

203-205 °C (lit.)

SMILES string

COc1cc2C=CC(=O)Oc2cc1O

InChI

1S/C10H8O4/c1-13-9-4-6-2-3-10(12)14-8(6)5-7(9)11/h2-5,11H,1H3

InChI key

RODXRVNMMDRFIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Scopoletin significantly increases activity of lipoprotein lipase in adipocytes by promoting transcription of the LPL gene. It also partially reverses inhibition of LPL by TNF-α.[1]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gui-Ping Zhang et al.
Journal of natural products, 72(7), 1265-1268 (2009-07-04)
Six new prenylated isoflavanones named sophoronols A-F (1-6), together with eight phenolic constituents, were isolated from the roots of Sophora mollis. Their structures and stereochemistry were established by 1D and 2D NMR techniques, especially HMBC and NOESY as well as
Soyoung Kim et al.
Journal of natural products, 69(12), 1769-1775 (2006-12-28)
Two new cyclopenta[b]benzofurans, aglaroxin A 1-O-acetate (2) and 3'-methoxyaglaroxin A 1-O-acetate (3), a new benzo[b]oxepine, 19,20-dehydroedulisone A (4), and five new cyclopenta[bc]benzopyrans, edulirin A (5), edulirin A 10-O-acetate (6), 19,20-dehydroedulirin A (7), isoedulirin A (8), and edulirin B (9), were
Jih-Jung Chen et al.
Journal of natural products, 71(2), 212-217 (2008-01-24)
Eight new compounds, including four new neolignans, (7' S,8' S)-bilagrewin ( 1), (7' S,8' S)-5-demethoxybilagrewin ( 2), (7' S,8' S)-5- O-demethyl-4'- O-methylbilagrewin ( 3), and (7' S,8' S)-nocomtal ( 4), a new coumarinolignan, (7' S,8' S)-4'- O-methylcleomiscosin D ( 5)
Shixin Deng et al.
Journal of natural products, 70(5), 859-862 (2007-03-24)
A phytochemical study of the fruits of noni (Morinda citrifolia) collected in Tahiti led to the isolation of two new lignans, (+)-3,4,3',4'-tetrahydroxy-9,7'alpha-epoxylignano-7 alpha,9'-lactone (1) and (+)-3,3'-bisdemethyltanegool (2), as well as seven known compounds, (-)-pinoresinol (3), (-)-3,3'-bisdemethylpinoresinol (4), quercetin (5), kaempferol
Maria E Riveiro et al.
Bioorganic & medicinal chemistry, 16(5), 2665-2675 (2007-12-07)
In the present study, we sought to establish the effect of diverse structural-related hydroxycoumarins on the proliferation, cytotoxicity, and induction of apoptosis in promonocytic leukemic cells (U-937). The dihydroxylated coumarins, 7,8-dihydroxy-coumarin and esculetin, induced DNA fragmentation as well as characteristic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service