Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

246522

Sigma-Aldrich

Allyl butyrate

99%

Synonym(s):

Allyl butanoate, Butanoic acid 2-propen-1-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH=CH2
CAS Number:
Molecular Weight:
128.17
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

99%

refractive index

n20/D 1.414 (lit.)

bp

44-45 °C/15 mmHg (lit.)

density

0.902 g/mL at 25 °C (lit.)

SMILES string

CCCC(=O)OCC=C

InChI

1S/C7H12O2/c1-3-5-7(8)9-6-4-2/h4H,2-3,5-6H2,1H3

InChI key

RMZIOVJHUJAAEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

107.6 °F - closed cup

flash_point_c

42 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service