246360
Chloroacetic anhydride
technical grade, ~90%
Synonym(s):
Monochloroacetic acid anhydride
About This Item
Recommended Products
grade
technical grade
vapor pressure
<0.75 mmHg ( 20 °C)
assay
~90%
bp
120-123 °C/20 mmHg (lit.)
mp
48-60 °C (lit.)
SMILES string
ClCC(=O)OC(=O)CCl
InChI
1S/C4H4Cl2O3/c5-1-3(7)9-4(8)2-6/h1-2H2
InChI key
PNVPNXKRAUBJGW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service