Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

243191

Sigma-Aldrich

Ethylamine

97%

Synonym(s):

Aminoethane, Monoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08
Beilstein/REAXYS Number:
505933
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor density

1.56 (15 °C, vs air)

vapor pressure

874 mmHg ( 20 °C)

assay

97%

autoignition temp.

721 °F

expl. lim.

14 %
3.5-14 %

bp

16.6 °C (lit.)

mp

-81 °C (lit.)

density

0.680 g/mL at 20 °C (lit.)
0.689 g/mL at 25 °C (lit.)

SMILES string

CCN

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3

Inchi Key

QUSNBJAOOMFDIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 301264-ALDRICH | Ethylamine 97%

Recommended products

Stainless steel needle valve Z146056 and hose adapter Z146838 is recommended.

hose barb

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

target_organs

Respiratory system

Storage Class

2A - Gases

wgk_germany

WGK 1

flash_point_f

-34.6 °F - closed cup

flash_point_c

-37 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Câline Christine et al.
Organic & biomolecular chemistry, 10(46), 9183-9190 (2012-10-23)
The synthesis of a phosphonated acyclic bifunctional chelate L* for the labeling of biomaterial is described. L* is based on a pyridine backbone, functionalized in ortho positions by aminomethyl-bis-methylphosphonic acids, and, in the para position, by a side chain containing
Lisa Vasicek et al.
Analytical chemistry, 81(19), 7876-7884 (2009-09-03)
Electron transfer dissociation (ETD) has proven to be a promising new ion activation method for proteomics applications due to its ability to generate c- and z-type fragment ions in comparison to the y- and b-type ions produced upon the more
Tracy A LeGreve et al.
The journal of physical chemistry. A, 113(2), 399-410 (2008-12-23)
Neutral serotonin-(H(2)O)(n) clusters with n = 1,2 have been studied under jet-cooled conditions using a combination of resonant two-photon ionization (R2PI), UV-UV hole-burning (UVHB), and resonant ion-dip infrared (RIDIR) spectroscopy. Serotonin (5-hydroxytryptamine, SERO) is a close analogue of tryptamine, differing
Song Yang et al.
Journal of chromatography. A, 1216(15), 3280-3289 (2009-03-10)
Complementary methods using liquid chromatography-tandem quadrupole mass spectrometry (LC-MS/MS) and comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry (GCxGC-TOF-MS) were developed and applied to determine targeted metabolites involved in central carbon metabolism [including tricarboxylic acid cycle, serine cycle, ethylmalonyl-coenzyme A (ethylmalonyl-CoA) pathway
Yuying Shuai et al.
Journal of the science of food and agriculture, 90(15), 2563-2567 (2010-08-26)
Theanine, a unique amino acid found almost exclusively in tea plants, has various favourable physiological and pharmacological functions in humans. Gamma-glutamyltranspeptidase (GGT, EC 2.3.2.2) is considered to be the most effective enzyme for the production of theanine. In fact, GGT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service