Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

240737

Sigma-Aldrich

2-Ethylhexanoic acid

≥99%

Synonym(s):

2-Ethylcaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CO2H
CAS Number:
Molecular Weight:
144.21
Beilstein/REAXYS Number:
1750468
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.98 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)
10 mmHg ( 115 °C)

assay

≥99%

autoignition temp.

699 °F

expl. lim.

1.04 %, 135 °F
8.64 %, 188 °F

refractive index

n20/D 1.425 (lit.)

bp

228 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)C(O)=O

InChI

1S/C8H16O2/c1-3-5-6-7(4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

OBETXYAYXDNJHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

237.2 °F - closed cup

flash_point_c

114 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Di Tommaso et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 40(1), 38-47 (2010-02-27)
Lyophilisation is a common procedure to increase the long-term stability of pharmaceutical formulations. Its applicability to polymeric micelles is usually an issue because of the aggregation of materials during freeze-drying steps. The feasibility of this process was studied on polymeric
Qian Zhang et al.
Nanoscale, 2(7), 1256-1259 (2010-07-22)
We report the synthesis of a series of monodispersed Bi-doped PbTe nanocrystals with tunable morphologies by using a doping precursor of bismuth(III) 2-ethylhexanoate. The as-synthesized Pb(1-x)Bi(x)Te (x = 0.005, 0.010, 0.015, 0.020) nanocrystals are characterized by X-ray diffraction, X-ray photoelectron
R Gaudin et al.
International archives of occupational and environmental health, 81(8), 959-966 (2007-12-22)
The aim of this study was to perform a biological monitoring survey of workers exposed to di(2-ethylhexyl)phthalate (DEHP) in a factory using polyvinyl chloride plastisols and to contribute additional occupational data of exposure particularly sparse in the industrial sectors where
I Svechnikova et al.
The Journal of endocrinology, 194(3), 603-609 (2007-09-01)
Phthalate esters are known to exert harmful effects on mammalian reproduction and fertility, but their potential adverse effects on the hormonal functions of the ovary have not yet been elucidated in detail. Here, we investigated the effects of di-(2-ethylhexyl) phthalate
S Elss et al.
Food additives and contaminants, 21(8), 811-814 (2004-09-17)
After simultaneous distillation-extraction (SDE) of commercial baby foods (n = 20) and fruit juices (n = 15) (among them 15 and eight products labelled 'organic', respectively) from 11 different suppliers, analyses performed by coupled capillary gas chromatography-mass spectrometry (GC-MS) revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service