Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

240087

Sigma-Aldrich

Dipropylamine

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)2NH
CAS Number:
Molecular Weight:
101.19
Beilstein/REAXYS Number:
505974
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥99%

refractive index

n20/D 1.4049 (lit.)

bp

105-110 °C (lit.)

density

0.738 g/mL at 25 °C (lit.)

SMILES string

CCCNCCC

InChI

1S/C6H15N/c1-3-5-7-6-4-2/h7H,3-6H2,1-2H3

InChI key

WEHWNAOGRSTTBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

44.6 °F - closed cup

flash_point_c

7 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service